Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Cngb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195413.1 | NP_001182342.1 | 5 - 6 | 565 | 94 | |
NM_145601.1 | NP_663576.1 | 5 - 6 | 565 | 94 | ||
GenBank mRNA | AK080705.1 | - | 5 - 6 | 552 | 94 | |
BC016201.1 | - | 5 - 6 | 565 | 4527347 | 94 | |
BC051415.1 | - | 4 - 5 | 519 | 94 | ||
HQ116386.1 | - | 5 - 6 | 565 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Ion Channel Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |