Search
Search

Commd7| Gene Symbol: Commd7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001195390.1 | NP_001182319.1 | 3 - 4 | 320 | 84 | |
| NM_133850.2 | NP_598611.2 | 2 - 3 | 245 | 84 | ||
| GenBank mRNA | AK008447.1 | - | 2 - 3 | 200 | 84 | |
| AK009274.1 | - | 2 - 3 | 220 | 84 | ||
| AK046582.1 | - | 2 - 3 | 195 | 84 | ||
| AK077349.1 | - | 2 - 3 | 186 | 84 | ||
| AK078774.1 | - | 2 - 3 | 204 | 84 | ||
| AK089926.1 | - | 2 - 3 | 188 | 84 | ||
| AK164805.1 | - | 2 - 3 | 154 | 84 | ||
| AK169037.1 | - | 3 - 4 | 267 | 84 | ||
| AK177311.1 | - | 3 - 4 | 169 | 84 | ||
| BC003310.1 | - | 2 - 3 | 178 | 84 | ||
| BC065794.1 | - | 2 - 3 | 146 | 84 | ||
| BC092215.1 | - | 2 - 3 | 162 | 6819383 | 84 | |
| BC094216.1 | - | 2 - 3 | 202 | 6820900 | 84 | |
Set Membership: |
|