Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250289_m1
          See other DBNDD1 GE Assays ›
          Gene Symbol:
          Dbndd1
          Gene Name:
          dysbindin (dystrobrevin binding protein 1) domain containing 1
          Gene Aliases:
          2810427I04Rik, D8Ertd590e
          Chromosome Location:
          Chr.8: 123504718 - 123518041 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250289_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Dbndd1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001170975.2 NP_001164446.2 1 - 2 218 69
          NM_001170976.1 NP_001164447.2 1 - 2 218 69
          NM_028146.4 NP_082422.3 1 - 2 218 69
          GenBank mRNA AK013173.1 - 1 - 2 117 69
          AK028187.1 - 1 - 2 155 69
          AK163434.1 - 1 - 2 218 69
          BC046622.1 - 1 - 2 153 6389902 69
          BQ554936.1 - 1 - 2 125 69

          Back To Top

          More Information


          Additional Information:

          This assay is in the 5'UTR region of the AK013173.1, AK028187.1, BC046622.1 transcripts.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Dbndd1

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240288 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240284 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240287 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997831 BC046622.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997831 NM_028146.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997831 AK013173.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997831 NM_001170975.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1997831 AK028187.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 453307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 811290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 448683 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70773 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 253263 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 627668 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928781 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 118552 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 171887 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P127160 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997831 BC046622.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997831 NM_028146.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997831 AK013173.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997831 NM_001170975.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1997831 AK028187.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240273 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240302 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240285 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240295 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240279 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240299 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240277 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240293 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240297 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240296 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240271 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240291 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240276 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26240287 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P127160 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06672493 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01850688 - -
          Whole Mouse Genome G4122A A_51_P497280 BC046622.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497280 NM_028146.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497280 AK013173.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P497280 NM_001170975.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P484289 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P132531 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P127160 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07233933 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07140816 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07331135 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07233931 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240284 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240286 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240289 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240287 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240290 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240285 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26240288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240270 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240336 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240242 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240221 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240326 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240219 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240334 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240314 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240293 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240330 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240337 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240271 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240277 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240296 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240262 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240229 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240265 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240343 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240307 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240345 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240340 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26240259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07204880 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240262 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07140816 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240283 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07268612 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06723588 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240227 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07080601 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240239 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240296 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07082870 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240298 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240221 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07233931 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240259 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07017984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240213 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06730969 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240219 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240299 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07082869 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26240229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07233933 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582555 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582553 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582554 - -
          Affymetrix HT MG-430 PM Array Plate 1449307_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582551 - -
          Mouse Genome 430 2.0 Array 1449307_PM_at - -
          Mouse Genome 430 2.0 Array 1449307_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72185_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 72185_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 NM_001170975.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 NM_001170975.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 NM_001170976.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 NM_001170976.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 NM_028146.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 NM_028146.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024136.mm.1 TC0800003102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 NM_001170975.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 NM_001170975.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 NM_001170976.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 NM_001170976.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 NM_028146.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 NM_028146.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024137.mm.1 TC0800003102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 NM_001170975.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 NM_001170975.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 NM_001170976.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 NM_001170976.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 NM_028146.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 NM_028146.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024135.mm.1 TC0800003102.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024131.mm.1 NM_001170976.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024131.mm.1 NM_001170976.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024131.mm.1 TC0800003102.mm.1 MTA TC
          Mouse Expression Set 430 1449307_at - -
          Mouse Expression Set 430 1449307_PM_at - -
          Mouse Genome 430A 2.0 Array 1449307_PM_at - -
          Mouse Genome 430A 2.0 Array 1449307_at - -
          Affymetrix HT Mouse Genome 430A Array 1449307_at - -
          Affymetrix HT Mouse Genome 430A Array 1449307_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 3170286 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003170286 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1227672 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1227672 - -
          mouse-6 v1.1 (Illumina) scl072185.1_8-S - -
          Illumina Sentrix mouse-6.v1 bead array 3170286 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl072185.1_8-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3170286 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1227672 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1227672 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl072185.1_8-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline