Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250425_m1
          See other RGS9 GE Assays ›
          Gene Symbol:
          Rgs9
          Gene Name:
          regulator of G-protein signaling 9
          Gene Aliases:
          RGS9-1, Rgs9-2
          Chromosome Location:
          Chr.11: 109225348 - 109298181 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250425_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Rgs9
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001165934.1 NP_001159406.1 1 - 2 249 59
          NM_011268.2 NP_035398.2 1 - 2 249 59
          GenBank mRNA AF011358.1 - 1 - 2 134 59
          AF125046.1 - 1 - 2 205 59
          AK085443.1 - 1 - 2 175 59
          AK138159.1 - 1 - 2 159 59
          AK164961.1 - 1 - 2 203 59
          BC129899.1 - 1 - 2 115 40111531 59
          BY277335.1 - 1 - 2 249 59

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100003986.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Rgs9

          Gene Ontology Categories:

          Function(s) Process(es)

          G-protein coupled receptor signaling pathway
          dopamine receptor signaling pathway
          visual perception
          regulation of G-protein coupled receptor protein signaling pathway
          negative regulation of signal transduction
          intracellular signal transduction
          termination of G-protein coupled receptor signaling pathway
          positive regulation of GTPase activity
          response to stimulus
          signal transducer activity
          GTPase activator activity
          protein binding
          protein complex binding

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1100037317.mm.1 NM_001165934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037317.mm.1 NM_001165934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037317.mm.1 NM_011268.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037317.mm.1 NM_011268.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037317.mm.1 TC1100003986.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100037320.mm.1 NM_001165934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037320.mm.1 NM_001165934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037320.mm.1 NM_011268.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037320.mm.1 NM_011268.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037320.mm.1 TC1100003986.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100037316.mm.1 NM_001165934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037316.mm.1 NM_001165934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037316.mm.1 NM_011268.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037316.mm.1 NM_011268.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037316.mm.1 TC1100003986.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100037322.mm.1 NM_001165934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037322.mm.1 NM_001165934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037322.mm.1 NM_011268.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037322.mm.1 NM_011268.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100037322.mm.1 TC1100003986.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline