Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01250796_m1
          See other CPM GE Assays ›
          Gene Symbol:
          Cpm
          Gene Name:
          carboxypeptidase M
          Gene Aliases:
          1110060I01Rik, 5730456K23Rik, AA589379, E030045M14Rik
          Chromosome Location:
          Chr.10: 117629500 - 117687352 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01250796_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cpm
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027468.1 NP_081744.1 2 - 3 194 63
          GenBank mRNA BC047389.1 - 1 - 2 180 63
          BC055073.1 - 2 - 3 199 63
          BC088735.1 - 2 - 3 199 63
          BC100404.1 - 2 - 3 188 30471377 63

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1000001366.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Protease Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          protease

          Gene Ontology Categories:

          Function(s) Process(es)

          proteolysis
          biological_process
          molecular_function
          carboxypeptidase activity
          metallocarboxypeptidase activity
          peptidase activity
          metallopeptidase activity
          zinc ion binding
          hydrolase activity
          metal ion binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482318 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482111 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482320 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482113 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 BC088735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 BC047389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 BC055073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 AK017670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 NM_027468.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 BC088735.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 BC047389.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 BC055073.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 AK017670.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P304397 NM_027468.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 748164 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 534582 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 936947 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 943942 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472791 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 864460 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 18995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AB086655.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AF477481.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK012041.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK017609.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK021216.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK028287.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK049163.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK076527.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK083015.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK213623.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 AK218896.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 BC013842.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 BC024485.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 BC048076.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 BC086955.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 916927 NM_026014.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 540281 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 472885 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 855511 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 468051 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 672350 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 355637 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 251111 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 319155 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 314559 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654803 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 538694 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 45062 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 98057 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 311246 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 858880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 42189 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 324253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 701553 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 352091 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 894221 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 606821 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 495396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 961261 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 377339 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 180047 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 300566 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952296 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 17439 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 177094 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 59290 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 133677 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 157576 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P164724 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P179785 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P155019 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 BC088735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 BC047389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 BC055073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 AK017670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 NM_027468.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 BC088735.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 BC047389.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 BC055073.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 AK017670.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P304397 NM_027468.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P179785 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P164724 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02229322 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P02229294 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P155019 - -
          Whole Mouse Genome G4122A A_51_P304397 BC088735.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 BC047389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 BC055073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 AK017670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 NM_027468.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 BC088735.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 BC047389.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 BC055073.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 AK017670.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P304397 NM_027468.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P155019 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P179785 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P164724 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482318 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482319 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482113 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482111 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P27482320 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482133 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482119 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482279 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482355 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482126 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08837529 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482120 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482339 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482116 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482141 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08374867 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482381 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08390278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08553406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482342 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482128 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482349 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08498026 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482362 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482122 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482345 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08662740 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482353 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482357 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08880034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482340 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08283111 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482099 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08837522 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482115 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482358 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482277 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482374 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482352 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482351 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482093 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08374868 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482347 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482127 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482131 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482134 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482136 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482244 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08801987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482382 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482271 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08390731 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482356 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27482125 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08821893 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10366551 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366549 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366548 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366547 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366550 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366552 - -
          Affymetrix Mouse Gene 1.0 ST Array 10366553 - -
          Affymetrix HT MG-430 PM Array Plate 1453009_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1429413_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10366546 - -
          Mouse Genome 430 2.0 Array 1429413_at - -
          Mouse Genome 430 2.0 Array 1458006_at - -
          Mouse Genome 430 2.0 Array 1429413_PM_at - -
          Mouse Genome 430 2.0 Array 1453009_PM_at - -
          Mouse Genome 430 2.0 Array 1453009_at - -
          Mouse Genome 430 2.0 Array 1458006_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70574_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70574_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1000010177.mm.1 NM_027468.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010177.mm.1 NM_027468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010177.mm.1 TC1000001366.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000010178.mm.1 NM_027468.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010178.mm.1 NM_027468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010178.mm.1 TC1000001366.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1000010174.mm.1 NM_027468.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010174.mm.1 NM_027468.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1000010174.mm.1 TC1000001366.mm.1 MTA TC
          Mouse Expression Set 430 1429413_PM_at - -
          Mouse Expression Set 430 1453009_PM_at - -
          Mouse Expression Set 430 1429413_at - -
          Mouse Expression Set 430 1453009_at - -
          Affymetrix HT Mouse Genome 430B Array 1429413_at - -
          Affymetrix HT Mouse Genome 430B Array 1453009_at - -
          Affymetrix HT Mouse Genome 430B Array 1429413_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1453009_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 514638 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5550066 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2629191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2629191 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2476452 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline