Search
Search

Ces2g| Gene Symbol: Ces2g | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_197999.2 | NP_932116.2 | 5 - 6 | 890 | 73 | |
| GenBank mRNA | AK146261.1 | - | 5 - 6 | 834 | 73 | |
| BC027185.1 | - | 5 - 6 | 868 | 4194872 | 73 | |
Set Membership: |
|