Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01251026_g1
          See other CBFB GE Assays ›
          Gene Symbol:
          Cbfb
          Gene Name:
          core binding factor beta
          Gene Aliases:
          AI893578, PEA2, PEBP2b, Pebp2, Pebpb2
          Chromosome Location:
          Chr.8: 105170674 - 105217989 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01251026_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Cbfb
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001161456.1 NP_001154928.1 1 - 2 384 68
          NM_001161457.1 NP_001154929.1 1 - 2 384 68
          NM_001161458.1 NP_001154930.1 1 - 2 384 68
          NM_022309.4 NP_071704.3 1 - 2 384 68
          GenBank mRNA AK089094.1 - 1 - 2 385 68
          BC006763.1 - 1 - 2 152 3481800 68
          BC026749.1 - 1 - 2 194 4016060 68
          BC040752.1 - 1 - 2 183 4016060 68
          D14570.1 - 1 - 2 314 68
          D14571.1 - 1 - 2 314 68
          D14572.1 - 1 - 2 314 68
          L03279.1 - 1 - 2 163 68
          L03305.1 - 1 - 2 122 68
          L03306.1 - 82 68

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800001204.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Cbfb

          Panther Classification:

          Molecular Function -

          DNA-binding transcription factor gene-specific transcriptional regulator

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          ossification
          osteoblast differentiation
          lymphocyte differentiation
          myeloid cell differentiation
          positive regulation of transcription from RNA polymerase II promoter
          cell maturation
          definitive hemopoiesis
          DNA binding
          transcription coactivator activity
          protein binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_001161456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_001161456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_001161457.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_001161457.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_001161458.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_001161458.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_022309.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 NM_022309.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009440.mm.1 TC0800001204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_001161456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_001161456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_001161457.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_001161457.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_001161458.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_001161458.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_022309.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 NM_022309.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009441.mm.1 TC0800001204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_001161456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_001161456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_001161457.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_001161457.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_001161458.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_001161458.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_022309.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 NM_022309.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009442.mm.1 TC0800001204.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_001161456.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_001161456.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_001161457.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_001161457.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_001161458.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_001161458.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_022309.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 NM_022309.4 Refseq
          Affymetrix Mouse Clariom S Array PSR0800009443.mm.1 TC0800001204.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 680652 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fbg7s:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline