Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252139_m1
          See other SLC16A5 GE Assays ›
          Gene Symbol:
          Slc16a5
          Gene Name:
          solute carrier family 16 (monocarboxylic acid transporters), member 5
          Gene Aliases:
          A130015N09Rik, MCT 5, MCT 6, MCT5
          Chromosome Location:
          Chr.11: 115462250 - 115474398 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252139_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Slc16a5
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001080934.1 NP_001074403.1 3 - 4 1294 70
          GenBank mRNA AK037416.1 - 2 - 3 1406 70
          BC172170.1 - 3 - 4 1309 70

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001895.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Transporter 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          biological_process
          plasma membrane lactate transport
          transmembrane transport
          molecular_function
          lactate transmembrane transporter activity
          symporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024054 BC172170.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024054 AK079757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024054 NM_001080934.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024046 BC053982.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024046 AK079757.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024046 BC052932.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2024046 NM_001080934.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 854435 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 256525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 389880 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435525 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 646911 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 150157 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 841504 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 568698 AK048816.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 58559 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024046 BC053982.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024046 AK079757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024046 BC052932.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024046 NM_001080934.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024054 BC172170.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024054 AK079757.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2024054 NM_001080934.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30027 BC053982.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30027 BC172170.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30027 AK079757.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30027 BC052932.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P30027 AK037416.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153721 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153671 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153619 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153694 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153733 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153659 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153725 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153782 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153658 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153734 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153788 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153790 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153640 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153786 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153716 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153772 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153630 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153720 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153625 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153668 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153689 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153738 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153784 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153722 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153633 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153663 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153731 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153779 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153718 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153681 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153638 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153714 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153656 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153792 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153746 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153621 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153744 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153710 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153775 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153712 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153777 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153673 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28153650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153718 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153659 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153698 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09637269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153648 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153617 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153656 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153650 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153713 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153733 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09182536 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153635 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09637278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153654 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153671 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09639137 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153619 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153694 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153727 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153673 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153721 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153621 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153682 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153725 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153699 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09516049 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09705376 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153686 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153665 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153638 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153631 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153614 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153637 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153646 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153624 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153666 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09154140 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153716 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153691 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153714 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09510370 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153705 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08989011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153634 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153712 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09318100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153625 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09509829 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153722 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153642 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153640 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153668 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153720 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09154139 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153678 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09029156 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153652 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09516054 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153676 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153664 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153708 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153700 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153627 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09630079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153663 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153658 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09705372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153681 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153643 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28153731 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382534 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382536 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382533 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382537 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382535 - -
          Affymetrix HT MG-430 PM Array Plate 1447225_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1434473_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382532 - -
          Mouse Genome 430 2.0 Array 1447225_PM_at - -
          Mouse Genome 430 2.0 Array 1434473_PM_at - -
          Mouse Genome 430 2.0 Array 1434473_at - -
          Mouse Genome 430 2.0 Array 1447225_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217316_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 217316_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100017833.mm.1 NM_001080934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017833.mm.1 NM_001080934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017833.mm.1 TC1100001895.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017831.mm.1 NM_001080934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017831.mm.1 NM_001080934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017831.mm.1 TC1100001895.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100017832.mm.1 NM_001080934.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017832.mm.1 NM_001080934.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100017832.mm.1 TC1100001895.mm.1 MTA TC
          Mouse Expression Set 430 1447225_PM_at - -
          Mouse Expression Set 430 1447225_at - -
          Mouse Expression Set 430 1434473_PM_at - -
          Mouse Expression Set 430 1434473_at - -
          Affymetrix HT Mouse Genome 430B Array 1447225_at - -
          Affymetrix HT Mouse Genome 430B Array 1434473_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434473_at - -
          Affymetrix HT Mouse Genome 430B Array 1447225_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 603808 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 102810446 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237289 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2461172 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline