Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252409_m1
          See other CCSAP GE Assays ›
          Gene Symbol:
          Ccsap
          Gene Name:
          centriole, cilia and spindle associated protein
          Gene Aliases:
          1700054N08Rik, AI853424
          Chromosome Location:
          Chr.8: 123839973 - 123860209 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252409_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ccsap
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_028536.1 NP_082812.1 2 - 3 734 72
          GenBank mRNA AK006786.1 - 1 - 2 126 72
          AK086180.1 - 1 - 2 439 72
          BC024705.1 - 1 - 2 391 3668303 72
          BC036300.1 - 2 - 3 734 4501054 72

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0800003160.mm.1

          Set Membership:

          Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Gene Ontology Categories:

          Function(s) Process(es)

          multicellular organism development
          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241996 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241993 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241992 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241998 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241999 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241995 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241997 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178152 BC024705.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178152 BC036300.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2178152 NM_028536.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 477741 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 239625 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404081 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 445862 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 425268 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 834719 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 248702 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 722531 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 729768 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 390253 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 684396 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 243995 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 628372 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 306608 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96532 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 111566 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P100585 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P166773 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178152 BC024705.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178152 BC036300.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2178152 NM_028536.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P01851159 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P166773 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P100585 - -
          Whole Mouse Genome G4122A A_52_P399814 BC024705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399814 BC036300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399814 NM_028536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399814 AK006786.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P399814 AK086180.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357094 BC024705.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357094 BC036300.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357094 NM_028536.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P357094 AK086180.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P166773 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P100585 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07428187 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07209554 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07216611 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06877824 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07209556 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07428200 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241998 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241992 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241994 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26242000 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241997 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241995 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241999 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241993 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26241996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242013 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241977 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241951 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241943 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241902 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241909 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241939 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241983 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241883 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241920 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241832 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241885 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242007 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241916 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241989 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241894 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242005 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241903 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241891 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242002 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241884 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241968 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241931 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242006 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241984 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242020 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241937 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241872 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241914 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241946 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241953 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241995 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241924 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241889 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241829 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241935 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241985 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241991 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241942 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241830 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242000 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241927 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241974 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241907 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242011 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241998 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241897 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241825 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241899 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241929 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241886 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241879 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241970 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241933 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241926 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241941 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241882 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241915 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241873 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242034 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241876 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241975 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241987 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241996 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241978 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241958 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241911 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241999 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241917 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241971 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241918 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241980 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242048 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241993 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26241831 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242019 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242017 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26242045 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241968 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241985 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241973 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242011 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242000 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241970 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242002 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241978 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241983 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07112504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07321530 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241960 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241993 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242016 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07404129 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07428200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242041 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241974 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06831623 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241987 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241971 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07001406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07254114 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241984 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241957 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241979 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07304004 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242006 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241977 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242034 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242029 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06877824 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242027 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241982 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07304007 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242014 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242013 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241958 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26241980 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07153203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P26242008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07428186 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07001409 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P07124025 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10582587 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582590 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582591 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582588 - -
          Affymetrix Mouse Gene 1.0 ST Array 10582589 - -
          Affymetrix Mouse Gene 1.0 ST Array 10576402 - -
          Affymetrix HT MG-430 PM Array Plate 1424796_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1451483_PM_s_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10582586 - -
          Affymetrix Mouse Gene 1.1 ST Array 10576401 - -
          Mouse Genome 430 2.0 Array 1424796_PM_at - -
          Mouse Genome 430 2.0 Array 1451483_PM_s_at - -
          Mouse Genome 430 2.0 Array 1424796_at - -
          Mouse Genome 430 2.0 Array 1451483_s_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73420_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 73420_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0800024243.mm.1 NM_028536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024243.mm.1 NM_028536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024243.mm.1 TC0800003160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024244.mm.1 NM_028536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024244.mm.1 NM_028536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024244.mm.1 TC0800003160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024245.mm.1 TC0800003160.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0800024247.mm.1 NM_028536.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024247.mm.1 NM_028536.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0800024247.mm.1 TC0800003160.mm.1 MTA TC
          Mouse Expression Set 430 1451483_PM_s_at - -
          Mouse Expression Set 430 1424796_at - -
          Mouse Expression Set 430 1424796_PM_at - -
          Mouse Expression Set 430 1451483_s_at - -
          Murine Genome U74v2 Set 171206_PM_f_at - -
          Murine Genome U74v2 Set 171206_f_at - -
          Mouse Genome 430A 2.0 Array 1451483_PM_s_at - -
          Mouse Genome 430A 2.0 Array 1451483_s_at - -
          Mouse Genome 430A 2.0 Array 1424796_at - -
          Mouse Genome 430A 2.0 Array 1424796_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1451483_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1451483_PM_s_at - -
          Affymetrix HT Mouse Genome 430A Array 1424796_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1424796_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 672649 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 60632 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1219407 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1219407 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline