Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Food and Beverage
    • Lab Solutions
    • Pharma and Biopharma
    • Real-Time PCR
    • Semiconductor Analysis
    • Clinical and Diagnostics
    • Digital Solutions
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • See all services
  • Help and Support
    • Order Help
    • Digital Solutions
    • Product Support
    • Technical Information
    • Training and Education
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252562_g1
          See other AANAT GE Assays ›
          Gene Symbol:
          Aanat
          Gene Name:
          arylalkylamine N-acetyltransferase
          Gene Aliases:
          AA-NAT, Nat-2, Nat4, Snat
          Chromosome Location:
          Chr.11: 116591721 - 116597680 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252562_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Aanat
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_009591.3 NP_033721.1 2 - 3 301 92
          NR_033223.1 2 - 3 301 92
          GenBank mRNA AB013358.1 - 1 - 2 157 92
          AF004108.1 - 2 - 3 301 92
          AF004110.1 - 2 - 3 301 92
          AF004111.1 - 1 - 2 213 92
          BC116967.1 - 2 - 3 272 40126286 92
          BC119139.1 - 2 - 3 272 8733888 92

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100001937.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Aanat

          Panther Classification:

          Molecular Function -

          acetyltransferase

          Gene Ontology Categories:

          Function(s) Process(es)

          N-terminal protein amino acid acetylation
          circadian rhythm
          response to zinc ion
          response to organic cyclic compound
          melatonin biosynthetic process
          response to insulin
          response to prostaglandin E
          response to copper ion
          rhythmic process
          cellular response to cAMP
          aralkylamine N-acetyltransferase activity
          arylamine N-acetyltransferase activity
          N-acetyltransferase activity
          transferase activity
          transferase activity, transferring acyl groups
          14-3-3 protein binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 NM_009591.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 NR_033223.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 AF004109.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 AF004110.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2158384 AF004111.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P497424 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 887632 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 481723 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 233782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 83162 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P137605 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 NM_009591.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 NR_033223.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 AF004109.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 AF004110.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2158384 AF004111.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P497424 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160267 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160260 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160254 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160249 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160263 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160251 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160259 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160261 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160269 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160274 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160272 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160270 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160265 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160256 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160258 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28160247 - -
          Whole Mouse Genome G4122A A_51_P295511 NM_009591.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295511 NR_033223.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295511 AF004108.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295511 AF004109.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P295511 AF004110.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P497424 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P131928 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372275 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09474817 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09372267 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160268 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160258 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160272 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160217 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160233 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160263 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160254 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160238 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160273 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160227 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160251 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160261 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160285 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160259 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160260 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160256 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160235 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160274 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28160240 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160273 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160268 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160235 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160287 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160230 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160245 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160233 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160252 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09260703 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160242 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160274 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160263 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160258 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160260 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160284 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160238 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09372270 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160281 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09653851 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09391849 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09114991 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09007243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160285 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160251 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160247 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09702208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160261 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160267 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09372269 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160272 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28160259 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10382820 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382828 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382818 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382827 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382826 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382825 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382819 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382829 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382824 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382817 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382822 - -
          Affymetrix Mouse Gene 1.0 ST Array 10382823 - -
          Affymetrix HT MG-430 PM Array Plate 1421666_PM_a_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10382816 - -
          Mouse Genome 430 2.0 Array 1421666_a_at - -
          Mouse Genome 430 2.0 Array 1421666_PM_a_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11298_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 11298_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100018408.mm.1 NM_009591.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018408.mm.1 NM_009591.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018408.mm.1 NR_033223.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018408.mm.1 NR_033223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018408.mm.1 TC1100001937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018411.mm.1 NM_009591.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018411.mm.1 NM_009591.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018411.mm.1 NR_033223.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018411.mm.1 NR_033223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018411.mm.1 TC1100001937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018410.mm.1 NR_033223.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018410.mm.1 NR_033223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018410.mm.1 TC1100001937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018412.mm.1 NM_009591.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018412.mm.1 NM_009591.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018412.mm.1 NR_033223.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018412.mm.1 NR_033223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018412.mm.1 TC1100001937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018413.mm.1 NM_009591.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018413.mm.1 NM_009591.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018413.mm.1 NR_033223.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018413.mm.1 NR_033223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018413.mm.1 TC1100001937.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1100018414.mm.1 NM_009591.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018414.mm.1 NM_009591.3 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018414.mm.1 NR_033223.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018414.mm.1 NR_033223.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100018414.mm.1 TC1100001937.mm.1 MTA TC
          Mouse Expression Set 430 1421666_PM_a_at - -
          Mouse Expression Set 430 1421666_a_at - -
          Murine Genome U74v2 Set 92511_at - -
          Murine Genome U74v2 Set 92512_g_at - -
          Murine Genome U74v2 Set 92512_PM_g_at - -
          Murine Genome U74v2 Set 92511_PM_at - -
          Mouse Genome 430A 2.0 Array 1421666_a_at - -
          Mouse Genome 430A 2.0 Array 1421666_PM_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421666_a_at - -
          Affymetrix HT Mouse Genome 430A Array 1421666_PM_a_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 622081 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 610609 - -
          Illumina mouse-6 v1.0 expression beadchip 610609 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0000610609 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2671661 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2671661 - -
          mouse-6 v1.1 (Illumina) scl40703.5.1_48-S - -
          Illumina Sentrix mouse-6.v1 bead array 610609 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl40703.5.1_48-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 610609 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2671661 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2671661 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline