Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
還沒有帳戶嗎 ? 立即註冊
  • 產品
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Western Blot Products
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Greener Products
    • 查看所有產品類別
  • 應用
    • 生物製程
    • 細胞培養和轉染
    • 食品和飲料
    • 實驗室解決方案
    • 製藥和生物製藥
    • 即時定量PCR
    • 半導體分析
    • 臨床和診斷
    • 數位解決方案
    • 分子光譜、同位素與元素分析
    • 查看所有應用與技術
  • 服務
    • 360° CDMO 和 CRO 服務
    • CDMO 服务
    • 临床试验 CRO 服务
    • 客製化服務
    • 金融與租賃服務
    • 儀器服務
    • 實驗室資訊學
    • 授權與商業供應
    • 培訓服務
    • 查看所有服務
  • 支援
    • 線上訂購協助
    • 數位解決方案
    • 產品支持
    • 技術相關資訊
    • 培訓與教育
    • 查看所有協助與支援
  • 熱門商品
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • 我們的服務對象
    • 生物技術
    • 生物製藥
    • CDMO
    • 實驗室診斷
    • 工業與應用科學
  • 特價商品
  • 聯絡我們
  • 快速訂購
  • 訂單狀態與貨物追蹤
  • 文件和證書
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • 訂單查詢
          • 快速訂購
          • 登入
            登入
            還沒有帳戶嗎 ? 立即註冊
            • 帳戶
            • 訂單
            • Connect: Lab, Data, Apps
            • 客製化產品與項目
            • 儀器管理
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252606_m1
          See other UNC13D GE Assays ›
          Gene Symbol:
          Unc13d
          Gene Name:
          unc-13 homolog D (C. elegans)
          Gene Aliases:
          2610108D09Rik, Jinx, Munc13-4, mFLJ00067
          Chromosome Location:
          Chr.11: 116062095 - 116078195 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252606_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Unc13d
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001009573.2 NP_001009573.2 13 - 14 1168 62
          GenBank mRNA AK220199.1 - 6 - 7 476 62
          BC141289.1 - 12 - 13 1080 9055901 62
          EF127645.1 - 12 - 13 1116 62

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1100004101.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Unc13d

          Gene Ontology Categories:

          Function(s) Process(es)

          granuloma formation
          germinal center formation
          exocytosis
          phagocytosis
          regulation of mast cell degranulation
          natural killer cell degranulation
          positive regulation of exocytosis
          defense response to virus
          positive regulation of substrate adhesion-dependent cell spreading
          positive regulation of regulated secretory pathway
          protein binding
          Rab GTPase binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976934 BC046519.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976928 AK220199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976928 NM_001009573.2 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 435919 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 727645 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 6730 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 101018 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 69642 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 286738 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 444316 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 204906 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 373325 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 81650 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 48238 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P133610 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P140912 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976928 AK220199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976928 NM_001009573.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976934 BC046519.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157231 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157236 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157257 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157243 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157242 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157247 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157237 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157250 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157252 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157246 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157255 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157240 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157245 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157253 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157241 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P28157234 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07504400 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P07504377 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P133610 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P140912 - -
          Whole Mouse Genome G4122A A_51_P512205 AK220199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512205 NM_001009573.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P512205 AK011827.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P133610 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P140912 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09180580 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09180615 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09180586 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09180610 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09180613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157143 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157212 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157222 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157205 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157145 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157177 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157313 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157320 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157138 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157302 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157171 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157312 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157246 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157166 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157139 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157201 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157197 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157237 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157190 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157322 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157137 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157305 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157136 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157310 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157211 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157173 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157298 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157193 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157152 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157255 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157208 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157247 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157250 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157318 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157252 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157196 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157245 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157135 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157174 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157243 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157150 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157224 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157297 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157141 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157149 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157253 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157182 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157186 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157295 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157161 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157257 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P28157300 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157225 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157222 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157211 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157257 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157223 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157231 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09180580 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09209939 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157208 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157234 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157250 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157206 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157216 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157236 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157246 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157248 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157228 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09180615 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09556827 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157205 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157255 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157212 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09180605 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157229 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09628147 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157220 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157243 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157241 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157237 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P28157245 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10393091 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393090 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393095 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393075 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393079 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393068 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393093 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393083 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393088 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393080 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393072 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393087 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393069 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393070 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393086 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393076 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393071 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393092 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393089 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393081 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393085 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393096 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393084 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393065 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393097 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393067 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393082 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393073 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393094 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393078 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393074 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393066 - -
          Affymetrix Mouse Gene 1.0 ST Array 10393077 - -
          Affymetrix HT MG-430 PM Array Plate 1428992_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10393064 - -
          Mouse Genome 430 2.0 Array 1428992_PM_at - -
          Mouse Genome 430 2.0 Array 1428992_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70450_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70450_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1100038621.mm.1 NM_001009573.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038621.mm.1 NM_001009573.2 Refseq
          Affymetrix Mouse Clariom S Array PSR1100038621.mm.1 TC1100004101.mm.1 MTA TC
          Mouse Expression Set 430 1428992_at - -
          Mouse Expression Set 430 1428992_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428992_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1428992_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 533689 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 104570736 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2502798 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2913913 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2502798 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2913913 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          產品訂購 Plus Icon Minus Icon
          • 訂單狀態
          • 訂購協助
          • 快速訂購
          • 現貨供應中心
          • 電子採購
          支持 Plus Icon Minus Icon
          • 協助與支持
          • 聯絡我們
          • 技術支持中心
          • 搜尋文件和證書
          • 回報網站問題
          資源 Plus Icon Minus Icon
          • 學習中心
          • 促銷活動
          • 活動和網路研討會
          • 社群媒體
          關於 Thermo Fisher Plus Icon Minus Icon
          • 關於我們 關於我們
          • 人才招募 人才招募
          • 投資者資訊 投資者資訊
          • 新聞 新聞
          • 企業社會責任 企業社會責任
          • 商標
          • 政策和聲明
          品牌 Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Taiwan flag icon
          Taiwan

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline