Search
Search
| Gene Symbol: Eea1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001932.3 | NP_001001932.1 | 21 - 22 | 3258 | 86 | |
| GenBank mRNA | BC075637.1 | - | 21 - 22 | 3171 | 30545051 | 86 |
Set Membership: |
|