Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01252771_s1
          See other OLFR412 GE Assays ›
          Gene Symbol:
          Olfr412
          Gene Name:
          olfactory receptor 412
          Gene Aliases:
          MOR127-5P
          Chromosome Location:
          Chr.11: 74364671 - 74365609 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01252771_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Olfr412
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001011851.1 NP_001011851.1 1 613 127

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Olfr412

          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          protein import into nucleus, translocation
          G-protein coupled receptor signaling pathway
          sensory perception of smell
          detection of chemical stimulus involved in sensory perception of smell
          olfactory receptor activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2022034 NM_001011851.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2022034 NM_001011851.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920063 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920078 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920087 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920074 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920088 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920095 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920090 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920094 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920086 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920053 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920068 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920067 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920081 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920084 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920082 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920070 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920057 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920061 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920072 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920055 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920077 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920060 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920079 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920075 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920092 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920062 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P27920097 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09062415 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09062403 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405044 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P09405042 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920078 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920067 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920070 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920045 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920068 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920075 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920053 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920109 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920090 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920087 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920092 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920094 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920110 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920119 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920077 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920062 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920046 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920055 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P27920072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920053 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09063393 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920096 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405042 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08992769 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09155191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920064 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920070 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920098 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920109 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09118278 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920107 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920092 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09062399 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09062403 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920087 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09149254 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P08994771 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09149253 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920104 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09149256 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920097 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09118276 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920067 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920055 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09405044 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920100 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920084 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920062 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P09062406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920101 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920105 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920112 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P27920060 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10378444 - -
          Affymetrix Mouse Gene 1.1 ST Array 10378443 - -
          Affymetrix Mouse Clariom S Array PSR1100009398.mm.1 NM_001011851.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009398.mm.1 NM_001011851.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1100009398.mm.1 TC1100001044.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 685314 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_3160736 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_3160736 - -
          mouse-6 v1.1 (Illumina) GI_58801459-S - -
          MouseRef-8 v2 Expression BeadChip ILMN_3160736 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_3160736 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) GI_58801459-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline