Search
Search

1700011L22Rik| Gene Symbol: 1700011L22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026315.1 | NP_080591.1 | 3 - 4 | 501 | 63 | |
| GenBank mRNA | AK005876.1 | - | 3 - 4 | 489 | 63 | |
Set Membership: |
|