Search
Search
| Gene Symbol: Irx6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301137.1 | NP_001288066.1 | 4 - 5 | 1458 | 142 | |
| NM_022428.3 | NP_071873.2 | 4 - 5 | 1458 | 142 | ||
| GenBank mRNA | AJ271055.1 | - | 4 - 5 | 722 | 142 | |
| AK035140.1 | - | 4 - 5 | 1461 | 142 | ||
| AK053729.1 | - | 4 - 5 | 1346 | 142 | ||
| BC036969.1 | - | 1 - 2 | 52 | 142 | ||
| BC130029.1 | - | 4 - 5 | 921 | 40140906 | 142 | |
Set Membership: |
|