| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100003031.mm.1 |
NM_030025.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003031.mm.1 |
NM_030025.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003031.mm.1 |
TC0100000390.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100003030.mm.1 |
NM_030025.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003030.mm.1 |
NM_030025.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003030.mm.1 |
TC0100000390.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100003032.mm.1 |
NM_030025.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003032.mm.1 |
NM_030025.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100003032.mm.1 |
TC0100000390.mm.1 |
MTA TC |