Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Pgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001163314.2 | NP_001156786.1 | 9 - 10 | 1188 | 116 | |
GenBank mRNA | AK041133.1 | - | 9 - 10 | 1189 | 116 | |
AK138171.1 | - | 9 - 10 | 1128 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |