Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01254729_m1
          See other SCGB2B26 GE Assays ›
          Gene Symbol:
          Scgb2b26
          Gene Name:
          secretoglobin, family 2B, member 26
          Gene Aliases:
          Abpbg26, Abpbg4, Abpg, C2d, Scgb2b4
          Chromosome Location:
          Chr.7: 33942997 - 33944985 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01254729_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 4,430.00
          Your Price
          Online offer:
          4430.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Scgb2b26
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_178308.2 NP_840093.1 2 - 3 254 101
          GenBank mRNA AY293282.1 - 1 - 2 164 101
          BC035222.1 - 2 - 3 240 4192901 101
          BC107292.1 - 2 - 3 242 40053055 101
          BC107293.1 - 2 - 3 242 40053056 101
          BC111446.1 - 2 - 3 254 40053210 101
          BC134362.1 - 2 - 3 260 40134659 101
          BC148302.2 - 2 - 3 260 40134662 101

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700002820.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Scgb2b26

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC134362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC148302.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC107292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC107293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC111446.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC134362.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC148302.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC107292.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC107293.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P58580 BC111446.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 882695 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 850301 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC134362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC148302.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC107292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC107293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC111446.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC134362.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC148302.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC107292.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC107293.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P58580 BC111446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC134362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC148302.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC107292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC107293.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC111446.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC134362.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC148302.2 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC107292.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC107293.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P58580 BC111446.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P137175 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P123510 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10562310 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562312 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562313 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562311 - -
          Affymetrix Mouse Gene 1.0 ST Array 10562309 - -
          Affymetrix Mouse Gene 1.1 ST Array 10562308 - -
          Affymetrix Mouse Clariom S Array PSR0700025179.mm.1 TC0700002820.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025179.mm.1 NM_178308.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025179.mm.1 NM_178308.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025183.mm.1 TC0700002820.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025183.mm.1 NM_178308.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025183.mm.1 NM_178308.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025180.mm.1 TC0700002820.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700025180.mm.1 NM_178308.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700025180.mm.1 NM_178308.2 Refseq
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4730594 - -
          MouseWG-6 v2 Expression BeadChip 5720736 - -
          Illumina mouse-6 v1.0 expression beadchip 5720736 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005720736 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2696136 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2599449 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2696136 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2599449 - -
          mouse-6 v1.1 (Illumina) scl31491.3.1_22-S - -
          Illumina Sentrix mouse-6.v1 bead array 5720736 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl31491.3.1_22-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5720736 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2771109 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2599449 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2696136 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2599449 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2771109 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2696136 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline