Search
Search

Fam117b| Gene Symbol: Fam117b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037725.3 | NP_001032814.1 | 3 - 4 | 990 | 62 | |
| GenBank mRNA | AK154812.1 | - | 3 - 4 | 992 | 62 | |
| BC080299.1 | - | 3 - 4 | 385 | 62 | ||
Set Membership: |
|