Search
Search

Ift74| Gene Symbol: Ift74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290568.1 | NP_001277497.1 | 11 - 12 | 1112 | 66 | |
| NM_026319.3 | NP_080595.2 | 11 - 12 | 1112 | 66 | ||
| GenBank mRNA | AK006492.1 | - | 2 - 3 | 219 | 66 | |
| AK011690.1 | - | 11 - 12 | 1083 | 66 | ||
| AK053369.1 | - | 11 - 12 | 1060 | 66 | ||
| BC005798.1 | - | 11 - 12 | 1082 | 66 | ||
| BC023760.1 | - | 11 - 12 | 1044 | 66 | ||
| BY706530.1 | - | 2 - 3 | 217 | 66 | ||
Set Membership: |
|