Search
Search

Ythdc2| Gene Symbol: Ythdc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163013.1 | NP_001156485.1 | 20 - 21 | 2777 | 59 | |
| GenBank mRNA | BC037178.1 | - | 7 - 8 | 799 | 59 | |
| BC040772.1 | - | 1 - 2 | 64 | 59 | ||
| BC072654.1 | - | 15 - 16 | 1821 | 59 | ||
| BC138263.1 | - | 20 - 21 | 2777 | 8861282 | 59 | |
| BC171951.1 | - | 20 - 21 | 2777 | 9054237 | 59 | |
Set Membership: |
|