| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048259 |
AK169724.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048259 |
BC047147.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048259 |
AK160454.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048259 |
AK167675.1 |
mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P2048259 |
NM_001177670.1 |
mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
945975 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
26981 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
420132 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
417491 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
864740 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
91558 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
890507 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
720757 |
AK051095.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
720757 |
NM_177894.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
947940 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
156695 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
594930 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
128792 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
664650 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
829435 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
277764 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
576935 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
360344 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
39137 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
637438 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
868015 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
436107 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
22291 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048259 |
AK169724.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048259 |
BC047147.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048259 |
AK160454.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048259 |
AK167675.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2048259 |
NM_001177670.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426014 |
BC027194.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426014 |
BC047147.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426014 |
AK160454.1 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426014 |
NM_146133.3 |
mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P426014 |
AK008434.1 |
mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344617 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344622 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344637 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344633 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344618 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344624 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344619 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344620 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344621 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344626 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344630 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344628 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344629 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344631 |
- |
- |
| Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P22344634 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04995508 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04995493 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00702844 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P00702806 |
- |
- |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P04995523 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P820985 |
- |
- |
| Whole Mouse Genome G4122A |
A_52_P588613 |
AK089547.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P350699 |
AK169724.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P350699 |
BC047147.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P350699 |
AK160454.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P350699 |
AK167675.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_52_P350699 |
NM_001177670.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P426014 |
BC027194.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P426014 |
BC047147.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P426014 |
AK160454.1 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P426014 |
NM_146133.3 |
mouse_transcriptome |
| Whole Mouse Genome G4122A |
A_51_P426014 |
AK008434.1 |
mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P100991 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P04995508 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00702844 |
- |
- |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_67_P00702806 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03150601 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03150605 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03150597 |
- |
- |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P03150595 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344680 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344778 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344702 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344740 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344752 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344762 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344642 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344734 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344617 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344756 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344750 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344713 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344796 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344782 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344736 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344732 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344687 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344622 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344629 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344777 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344644 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344634 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344792 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344789 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344626 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344624 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344628 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344733 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344705 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344620 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344769 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344738 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344653 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344794 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344619 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344744 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344712 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344747 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344716 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344745 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344699 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344788 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344701 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344714 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344676 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344637 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344722 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344723 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344779 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344753 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344692 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344691 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344673 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344639 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344670 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344781 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344785 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344655 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344755 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344666 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344770 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344684 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344737 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344773 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344677 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344703 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344742 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344638 |
- |
- |
| Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P22344695 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02441318 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344683 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344639 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02731503 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344680 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02618397 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03108103 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344622 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344687 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344658 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344677 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344690 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344673 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344628 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02687878 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344631 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344745 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344657 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344619 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344655 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344689 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344734 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02870661 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344742 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344616 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344740 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344629 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344617 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344701 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344653 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02536132 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344620 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02870662 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344642 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344647 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344684 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02495187 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344624 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344703 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344727 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344702 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344646 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344644 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344679 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344676 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344618 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02641921 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02641922 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344637 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03062959 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344726 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02721179 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P02687882 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344634 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344626 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344747 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344670 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344705 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344645 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344638 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03062958 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P03081809 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344708 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344728 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344744 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P22344686 |
- |
- |