Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257226_m1
          See other PSG29 GE Assays ›
          Gene Symbol:
          Psg29
          Gene Name:
          pregnancy-specific glycoprotein 29
          Gene Aliases:
          AA102956, AA409969, cea17
          Chromosome Location:
          Chr.7: 17203477 - 17215756 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257226_m1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price 435.00
          Your Price
          Online offer: 408.65
          408.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Psg29
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_054064.3 NP_473405.1 4 - 5 1326 118
          GenBank mRNA AK141330.1 - 4 - 5 1327 118
          BC114207.1 - 4 - 5 1303 30083585 118

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000252.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Mus musculus 1700 Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Psg29

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067787 NM_054064.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067787 BC114207.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2067787 CK336734.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 915980 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 549381 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 260868 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 214274 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 618471 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 181903 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 715539 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 170646 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067787 NM_054064.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067787 BC114207.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2067787 CK336734.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985172 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985154 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985162 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985145 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985176 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985159 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985178 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985169 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985181 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985164 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985151 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985175 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985170 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985165 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985157 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985146 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985156 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985160 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985158 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985179 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985150 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985167 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985147 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985182 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985163 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24985173 - -
          Whole Mouse Genome G4122A A_51_P155783 NM_054064.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155783 AK141330.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P155783 BC114207.1 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985160 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985179 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985185 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985225 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985162 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985236 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985218 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985157 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985146 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985151 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985226 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985194 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985189 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985210 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985241 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985156 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985183 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985167 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985228 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985230 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985154 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985188 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985175 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985184 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985198 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985178 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985199 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985220 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985204 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985223 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985216 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985192 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985231 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985215 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985159 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985169 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985214 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985172 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985176 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985206 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985164 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985163 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985234 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985147 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985232 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985239 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985181 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985187 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985203 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985213 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985191 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985209 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24985158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985189 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985178 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985198 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985210 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985165 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985192 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06616265 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919916 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985169 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985191 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985157 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985185 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914379 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919918 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919919 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919904 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985215 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985159 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985200 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985170 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985182 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985154 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06246028 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985187 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914377 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985194 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985196 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919929 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985160 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05919937 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985209 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985203 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985183 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985175 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985151 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06357018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985145 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985167 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985207 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985214 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985164 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914372 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05914361 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985199 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985176 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985150 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985148 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985204 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985179 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24985158 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06518317 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550429 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550426 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550427 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550431 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550432 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550425 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550430 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550428 - -
          Affymetrix Mouse Gene 1.1 ST Array 10550424 - -
          Affymetrix Mouse Clariom S Array PSR0700001883.mm.1 TC0700000252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001883.mm.1 NM_054064.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001883.mm.1 NM_054064.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001882.mm.1 TC0700000252.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001882.mm.1 NM_054064.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001882.mm.1 NM_054064.3 Refseq
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 476991 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1249265 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-zdkzh:80/100.66.76.145:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline