Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257351_g1
          See other MCL1 GE Assays ›
          Gene Symbol:
          Mcl1
          Gene Name:
          myeloid cell leukemia sequence 1
          Gene Aliases:
          AW556805, Mcl-1
          Chromosome Location:
          Chr.3: 95658368 - 95663179 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257351_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Mcl1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_008562.3 NP_032588.1 1 - 2 771 127
          GenBank mRNA AK159504.1 - 1 - 2 683 127
          BC003839.1 - 1 - 2 700 3491015 127
          BC005427.1 - 1 - 2 693 3493739 127
          BC021638.1 - 1 - 2 710 4190996 127
          GU182318.1 - 2 - 3 497 127
          U35623.1 - 1 - 2 683 127

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0300000910.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length greater than or equal to 101 Apoptosis Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Mcl1

          Gene Ontology Categories:

          Function(s) Process(es)

          apoptotic process
          multicellular organism development
          intrinsic apoptotic signaling pathway in response to DNA damage
          apoptotic mitochondrial changes
          cell differentiation
          response to cytokine
          regulation of apoptotic process
          negative regulation of apoptotic process
          extrinsic apoptotic signaling pathway in absence of ligand
          positive regulation of oxidative stress-induced neuron intrinsic apoptotic signaling pathway
          negative regulation of anoikis
          regulation of response to DNA damage stimulus
          negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
          negative regulation of intrinsic apoptotic signaling pathway
          protein binding
          protein homodimerization activity
          protein heterodimerization activity
          protein dimerization activity
          BH domain binding
          BH3 domain binding

          MicroArray Probes:

          Affymetrix ABI

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR0300007246.mm.1 NM_008562.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007246.mm.1 NM_008562.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007246.mm.1 TC0300000910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007248.mm.1 NM_008562.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007248.mm.1 NM_008562.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007248.mm.1 TC0300000910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007247.mm.1 NM_008562.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007247.mm.1 NM_008562.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007247.mm.1 TC0300000910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007242.mm.1 NM_008562.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007242.mm.1 NM_008562.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007242.mm.1 TC0300000910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007249.mm.1 NM_008562.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007249.mm.1 NM_008562.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007249.mm.1 TC0300000910.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0300007245.mm.1 NM_008562.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007245.mm.1 NM_008562.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0300007245.mm.1 TC0300000910.mm.1 MTA TC
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 665192 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-67655d8755-x75fr:80/100.66.77.4:80.
          git-commit: dddaa802cd395f65bf1581942d0e97a089c38f41
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.46.0-Offline