Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257500_g1
          See other FBXO6 GE Assays ›
          Gene Symbol:
          Fbxo6
          Gene Name:
          F-box protein 6
          Gene Aliases:
          AA408845, FBG2, Fbs2, Fbx6b, Fbxo6b
          Chromosome Location:
          Chr.4: 148145716 - 148152135 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257500_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fbxo6
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001163704.1 NP_001157176.1 4 - 5 822 62
          NM_001163705.1 NP_001157177.1 4 - 5 606 62
          NM_001163706.1 NP_001157178.1 4 - 5 598 62
          NM_001163707.1 NP_001157179.1 4 - 5 633 62
          NM_015797.4 NP_056612.1 4 - 5 1348 62
          GenBank mRNA AF176526.1 - 4 - 5 721 62
          AK002840.1 - 4 - 5 560 62
          AK144715.1 - 4 - 5 759 62
          AK150612.1 - 4 - 5 790 62
          AK152888.1 - 4 - 5 672 62
          AK165867.1 - 4 - 5 1333 62
          AK170743.1 - 4 - 5 599 62
          AK171483.1 - 4 - 5 599 62
          BC017512.1 - 4 - 5 618 4485218 62

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fbxo6

          Gene Ontology Categories:

          Function(s) Process(es)

          DNA repair
          glycoprotein catabolic process
          cellular response to DNA damage stimulus
          response to unfolded protein
          ER-associated ubiquitin-dependent protein catabolic process
          SCF-dependent proteasomal ubiquitin-dependent protein catabolic process
          glycoprotein binding
          protein binding
          carbohydrate binding
          ubiquitin protein ligase activity

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392699 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392701 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392700 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392702 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074291 BC017512.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074291 AK150612.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074291 AK165867.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074291 AK144715.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2074291 NM_001163707.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 242346 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 654644 AK035857.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70793 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905135 AF305614.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905135 AK090082.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905135 AK152288.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905135 AK167036.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905135 BC003907.1 genbank_mrna_mouse
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 905135 NM_025433.3 refseq_mrna_mouse_NM
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 71897 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P136878 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074291 BC017512.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074291 AK150612.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074291 AK165867.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074291 AK144715.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2074291 NM_001163707.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392701 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392705 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392707 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392688 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392691 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392693 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392700 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392690 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392699 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392686 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392695 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392703 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392692 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23392698 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P136878 - -
          Whole Mouse Genome G4122A A_52_P557923 AK170743.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557923 AK171483.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557923 BC017512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557923 AK150612.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P557923 AK152888.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228846 BC017512.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P228846 AF176526.1 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P136458 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P136878 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03945829 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03945824 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03445818 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03945827 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392699 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392700 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392701 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P23392702 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392747 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392698 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392611 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392762 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392678 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392650 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392610 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392622 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392676 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392615 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392703 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392688 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392745 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392737 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392669 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392742 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392629 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392637 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392617 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392691 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392627 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392764 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392672 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392646 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392693 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392616 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392657 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392758 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392626 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392699 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392618 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392606 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392639 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392649 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392766 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392589 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392696 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392642 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392644 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392599 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392645 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392686 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392635 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392590 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392666 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392700 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392680 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392707 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392683 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392660 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392652 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392631 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392761 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392665 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392682 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392752 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392624 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392648 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392748 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392751 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392736 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392596 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392749 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392754 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392613 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392743 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392593 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392759 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392732 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392662 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392750 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392701 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392729 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392654 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392609 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392603 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392674 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392727 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392690 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392632 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392608 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392741 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23392653 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392696 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392702 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392669 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392655 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04055053 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392707 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392674 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392687 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392657 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392683 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392698 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392678 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392670 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392699 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392652 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392680 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392684 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392691 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392695 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392672 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392676 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392660 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03569668 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03321816 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392693 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392663 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392662 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392690 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03445818 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392666 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03945829 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23392665 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03321821 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10518479 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518481 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518478 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518475 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518474 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518476 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518477 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518482 - -
          Affymetrix Mouse Gene 1.0 ST Array 10518480 - -
          Affymetrix HT MG-430 PM Array Plate 1447671_PM_x_at - -
          Affymetrix HT MG-430 PM Array Plate 1417501_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10518473 - -
          Mouse Genome 430 2.0 Array 1447671_x_at - -
          Mouse Genome 430 2.0 Array 1447671_PM_x_at - -
          Mouse Genome 430 2.0 Array 1417501_at - -
          Mouse Genome 430 2.0 Array 1417501_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50762_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 50762_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 TC0400004014.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163704.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163704.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163705.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163705.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163706.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163706.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163707.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_001163707.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_015797.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400033185.mm.1 NM_015797.4 Refseq
          Mouse Expression Set 430 1417501_PM_at - -
          Mouse Expression Set 430 1417501_at - -
          Mouse Expression Set 430 1447671_x_at - -
          Mouse Expression Set 430 1447671_PM_x_at - -
          Murine Genome U74v2 Set 95014_at - -
          Murine Genome U74v2 Set 162050_at - -
          Murine Genome U74v2 Set 162050_PM_at - -
          Murine Genome U74v2 Set 95014_PM_at - -
          Mouse Genome 430A 2.0 Array 1417501_PM_at - -
          Mouse Genome 430A 2.0 Array 1417501_at - -
          Affymetrix HT Mouse Genome 430A Array 1417501_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1417501_at - -
          Affymetrix HT Mouse Genome 430B Array 1447671_x_at - -
          Affymetrix HT Mouse Genome 430B Array 1447671_PM_x_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 5690692 - -
          Illumina mouse-6 v1.0 expression beadchip 5690692 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0005690692 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2660175 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2660175 - -
          mouse-6 v1.1 (Illumina) scl23523.7.1_20-S - -
          Illumina Sentrix mouse-6.v1 bead array 5690692 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl23523.7.1_20-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690692 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2660175 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2660175 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl23523.7.1_20-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline