Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01257720_m1
          See other PIAS2 GE Assays ›
          Gene Symbol:
          Pias2
          Gene Name:
          protein inhibitor of activated STAT 2
          Gene Aliases:
          6330408K17Rik, AI462206, ARIP3, AU018068, Dib, Miz1, PIASxalpha, PIASxb, PIASxbeta, SIZ2
          Chromosome Location:
          Chr.18: 77065208 - 77155710 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01257720_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 284.00
          Your Price
          Online offer: 266.65
          266.65
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Pias2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001164167.1 NP_001157639.1 11 - 12 1695 89
          NM_001164168.1 NP_001157640.1 11 - 12 1711 89
          NM_001164169.1 NP_001157641.1 11 - 12 1695 89
          NM_001164170.1 NP_001157642.1 11 - 12 1711 89
          NM_008602.4 NP_032628.3 11 - 12 1695 89
          GenBank mRNA AF039567.1 - 10 - 11 1140 89
          AF201391.1 - 11 - 12 1622 89
          AK014871.1 - 11 - 12 1686 89
          AK029716.1 - 11 - 12 1696 89
          AK032463.1 - 1 - 2 130 89
          AK078813.1 - 11 - 12 1665 89
          AK086653.1 - 11 - 12 1627 89
          AK149142.1 - 12 - 13 1734 89
          BC005596.1 - 11 - 12 1572 3969546 89
          BC034711.1 - 11 - 12 1701 2631738 89

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800000825.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Mus musculus Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Pias2

          Gene Ontology Categories:

          Function(s) Process(es)

          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          protein sumoylation
          androgen receptor signaling pathway
          negative regulation of sequence-specific DNA binding transcription factor activity
          regulation of osteoblast differentiation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of dendrite morphogenesis
          regulation of androgen receptor signaling pathway
          negative regulation of androgen receptor signaling pathway
          DNA binding
          protein binding
          transcription factor binding
          zinc ion binding
          UDP-N-acetylmuramoylalanyl-D-glutamyl-2,6-diaminopimelate-D-alanyl-D-alanine ligase activity
          ligase activity
          ribosomal S6-glutamic acid ligase activity
          SUMO transferase activity
          protein domain specific binding
          estrogen receptor binding
          ubiquitin protein ligase binding
          glucocorticoid receptor binding
          coenzyme F420-0 gamma-glutamyl ligase activity
          coenzyme F420-2 alpha-glutamyl ligase activity
          metal ion binding
          androgen receptor binding
          SUMO ligase activity
          protein-glycine ligase activity
          protein-glycine ligase activity, initiating
          protein-glycine ligase activity, elongating
          tubulin-glycine ligase activity
          protein-glutamic acid ligase activity
          tubulin-glutamic acid ligase activity

          MicroArray Probes:

          Affymetrix

          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 NM_001164167.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 NM_001164167.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 NM_001164168.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 NM_001164168.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 NM_008602.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 NM_008602.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005990.mm.1 TC1800000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164167.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164167.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164168.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164168.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164169.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164169.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164170.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_001164170.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_008602.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 NM_008602.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005987.mm.1 TC1800000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005992.mm.1 NM_001164168.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005992.mm.1 NM_001164168.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005992.mm.1 NM_008602.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005992.mm.1 NM_008602.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005992.mm.1 TC1800000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 NM_001164167.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 NM_001164167.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 NM_001164168.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 NM_001164168.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 NM_008602.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 NM_008602.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005994.mm.1 TC1800000825.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 NM_001164167.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 NM_001164167.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 NM_001164168.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 NM_001164168.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 NM_008602.4.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 NM_008602.4 Refseq
          Affymetrix Mouse Clariom S Array PSR1800005993.mm.1 TC1800000825.mm.1 MTA TC

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Spain flag icon
          Spain

          TEST

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline