Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01258933_s1
          See other FV1 GE Assays ›
          Gene Symbol:
          Fv1
          Gene Name:
          Friend virus susceptibility 1
          Gene Aliases:
          Fv-1, Rv-1, Rv1
          Chromosome Location:
          Chr.4: 147868979 - 147870358 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01258933_s1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Fv1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_010244.3 NP_034374.2 1 716 79

          Back To Top

          More Information


          Additional Information:

          Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Mus musculus Inventoried Both primers and probe map within a single exon

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Fv1

          Gene Ontology Categories:

          Function(s)

          response to virus

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P421223 NM_010244.3 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P421223 NM_010244.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 928312 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P421223 NM_010244.3 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P421223 NM_010244.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23391046 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23391044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23391043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23391041 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P23391045 - -
          Whole Mouse Genome G4122A A_51_P421223 NM_010244.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P421223 NM_010244.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P674524 NM_010244.3 mouse_transcriptome
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P23391039 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23391040 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03865586 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10510255 - -
          Affymetrix HT MG-430 PM Array Plate 1422279_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10510254 - -
          Mouse Genome 430 2.0 Array 1422279_at - -
          Mouse Genome 430 2.0 Array 1422279_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14349_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 14349_at - -
          Affymetrix Mouse Clariom S Array PSR0400015631.mm.1 NM_010244.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015631.mm.1 NM_010244.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015631.mm.1 TC0400001918.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400015630.mm.1 NM_010244.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015630.mm.1 NM_010244.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400015630.mm.1 TC0400001918.mm.1 MTA TC
          Mouse Expression Set 430 1422279_PM_at - -
          Mouse Expression Set 430 1422279_at - -
          Murine Genome U74v2 Set 99807_r_at - -
          Murine Genome U74v2 Set 99807_PM_r_at - -
          Mouse Genome 430A 2.0 Array 1422279_at - -
          Mouse Genome 430A 2.0 Array 1422279_PM_at - -
          Affymetrix HT Mouse Genome 430A Array 1422279_at - -
          Affymetrix HT Mouse Genome 430A Array 1422279_PM_at - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.0 expression beadchip 2230102 - -
          Illumina mouse-6 v1.0 expression beadchip 106110072 - -
          Illumina mouse-6 v1.0 expression beadchip 3450746 - -
          Illumina mouse-6 v1.0 expression beadchip 1450110 - -
          Illumina mouse-6 v1.0 expression beadchip 1770402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001770402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0003450746 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0001450110 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002230102 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1220594 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1237595 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2736941 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1232454 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1232454 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1237595 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2736941 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1220594 - -
          mouse-6 v1.1 (Illumina) scl0014349.2_95-S - -
          mouse-6 v1.1 (Illumina) GI_34328175-S - -
          mouse-6 v1.1 (Illumina) scl24713.1_10-S - -
          mouse-6 v1.1 (Illumina) scl0014349.1_36-S - -
          mouse-6 v1.1 (Illumina) scl0014349.2_37-S - -
          Illumina Sentrix mouse-6.v1 bead array 1450110 - -
          Illumina Sentrix mouse-6.v1 bead array 2230102 - -
          Illumina Sentrix mouse-6.v1 bead array 1770402 - -
          Illumina Sentrix mouse-6.v1 bead array 3450746 - -
          Illumina Sentrix mouse-6.v1 bead array 106110072 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl24713.1_10-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014349.2_37-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014349.1_36-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0014349.2_95-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1770402 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 3450746 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 106110072 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2230102 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 1450110 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1237595 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1220594 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2736941 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1220594 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1257107 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1237595 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2736941 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1232454 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-green-b49b87d85-hznln:80/100.66.79.29:80.
          git-commit: 5b8c860b7cdb41e9cfe07630520f6b51e109d38e
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.47.0-Offline