Search
Search

Ptprn| Gene Symbol: Ptprn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008985.2 | NP_033011.2 | 4 - 5 | 471 | 57 | |
| GenBank mRNA | AF332083.1 | - | 1 - 2 | 75 | 57 | |
| AF332084.1 | - | 1 - 2 | 75 | 57 | ||
| AK041296.1 | - | 4 - 5 | 477 | 57 | ||
| AK076223.1 | - | 4 - 5 | 450 | 57 | ||
| AK139398.1 | - | 4 - 5 | 471 | 57 | ||
| AK163141.1 | - | 4 - 5 | 447 | 57 | ||
| AK220467.1 | - | 4 - 5 | 429 | 57 | ||
| BM950464.1 | - | 2 - 3 | 137 | 57 | ||
| DQ832283.1 | - | 4 - 5 | 448 | 57 | ||
| U11812.1 | - | 4 - 5 | 395 | 57 | ||
| X74438.1 | - | 4 - 5 | 488 | 57 | ||
Set Membership: |
|