Search
Search

Zfp37| Gene Symbol: Zfp37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290350.1 | NP_001277279.1 | 3 - 4 | 446 | 57 | |
| NM_001290351.1 | NP_001277280.1 | 3 - 4 | 437 | 57 | ||
| NM_001290353.1 | NP_001277282.1 | 3 - 4 | 344 | 57 | ||
| NM_009554.4 | NP_033580.3 | 3 - 4 | 434 | 57 | ||
| GenBank mRNA | AK032469.1 | - | 3 - 4 | 316 | 57 | |
| BC024863.1 | - | 1 - 2 | 57 | 57 | ||
| BC063757.1 | - | 3 - 4 | 318 | 6331110 | 57 | |
| CF915126.1 | - | 3 - 4 | 344 | 57 | ||
| CO430987.1 | - | 3 - 4 | 316 | 57 | ||
| CO434318.1 | - | 3 - 4 | 271 | 57 | ||
| X64413.1 | - | 3 - 4 | 240 | 57 | ||
| X89264.1 | - | 4 - 5 | 346 | 57 | ||
Set Membership: |
|