Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01259306_m1
          See other PRPF4 GE Assays ›
          Gene Symbol:
          Prpf4
          Gene Name:
          PRP4 pre-mRNA processing factor 4 homolog (yeast)
          Gene Aliases:
          1600015H11Rik, AI874830, AW047464, bN189G18.1
          Chromosome Location:
          Chr.4: 62408767 - 62426990 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01259306_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Prpf4
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_027297.3 NP_081573.1 10 - 11 1112 65
          GenBank mRNA AK005464.1 - 10 - 11 1091 65
          AK085174.1 - 10 - 11 1076 65
          AK089838.1 - 10 - 11 1064 65
          AK090114.1 - 10 - 11 1089 65
          AK166912.1 - 10 - 11 1098 65
          AK167216.1 - 10 - 11 1090 65
          AK169532.1 - 10 - 11 1075 65
          BC051639.1 - 6 - 7 526 65
          BC106089.1 - 10 - 11 1074 30715403 65
          BC125529.1 - 10 - 11 1063 40130044 65
          BC125531.1 - 10 - 11 1063 40130046 65

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400000700.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Prpf4

          Panther Classification:

          Molecular Function -

          RNA splicing factor

          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          mRNA processing
          RNA splicing
          molecular_function
          U6 snRNA binding
          U4 snRNA binding

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964279 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964278 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2115003 NM_027297.3 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 806879 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 653643 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 222257 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 879209 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 922469 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 516244 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 127623 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270307 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 120784 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 229767 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 84203 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 661614 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 420425 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 802112 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 132782 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 152300 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2115003 NM_027297.3 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964288 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964292 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964298 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964303 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964280 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964290 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964307 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964283 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964294 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964305 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964286 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964281 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964284 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964278 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22964300 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05257394 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05257343 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00886186 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P00886191 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05257365 - -
          Whole Mouse Genome G4122A A_51_P193852 BC051639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193852 AK169532.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193852 AK167216.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193852 NM_027297.3 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P193852 AK085174.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536082 BC125529.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536082 BC125531.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536082 BC051639.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536082 BC106089.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P536082 AK169532.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05257365 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_67_P05257394 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03720524 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03720502 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964278 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P22964279 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964290 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964276 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964401 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964429 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964280 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964291 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964278 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964410 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964427 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964376 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964453 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964441 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964447 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964348 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964395 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964319 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964407 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964308 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964281 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964423 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964382 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964439 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964444 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964335 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964402 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964311 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964406 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964331 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964362 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964418 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964413 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964364 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964303 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964432 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964370 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964339 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964386 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964374 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964405 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964316 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964323 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964292 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964367 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964344 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964359 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964287 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964397 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964377 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964372 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964357 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964392 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964283 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964442 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964431 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964294 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964438 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964387 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964415 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964373 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964365 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964381 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964327 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964329 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964368 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964342 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964321 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964424 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964379 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964346 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964389 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964411 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964420 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964414 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964393 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964288 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964433 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964325 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964351 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964333 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964399 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964446 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964301 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964317 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964284 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22964350 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516311 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964294 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04086391 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516326 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03739563 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516317 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03382752 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964292 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964287 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964351 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964300 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03720525 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964344 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278719 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03720503 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278717 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03373556 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03720520 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964291 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P04086387 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278728 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964280 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516330 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03720518 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964352 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964323 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278734 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964308 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03516310 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964278 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03278723 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964314 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964346 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964298 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964296 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P03780477 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964283 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964295 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964303 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964321 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964288 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964305 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P22964317 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10505298 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505289 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505287 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505292 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505291 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505297 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505286 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505288 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505293 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505294 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505283 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505284 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505290 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505296 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505295 - -
          Affymetrix Mouse Gene 1.0 ST Array 10505285 - -
          Affymetrix HT MG-430 PM Array Plate 1429724_PM_at - -
          Affymetrix HT MG-430 PM Array Plate 1438481_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10505282 - -
          Mouse Genome 430 2.0 Array 1429724_PM_at - -
          Mouse Genome 430 2.0 Array 1429724_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70052_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 70052_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400004920.mm.1 NM_027297.3.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004920.mm.1 NM_027297.3 Refseq
          Affymetrix Mouse Clariom S Array PSR0400004920.mm.1 TC0400000700.mm.1 MTA TC
          Mouse Expression Set 430 1429724_PM_at - -
          Mouse Expression Set 430 1429724_at - -
          Affymetrix HT Mouse Genome 430B Array 1438481_at - -
          Affymetrix HT Mouse Genome 430B Array 1429724_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438481_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1429724_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 426518 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 2970364 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1253680 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2700079 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2700079 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1253680 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-7d94cb4b65-jwmpg:80/100.66.76.145:80.
          git-commit: c9e08c96761173abe34e68f880379696776a4827
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.44.1-2026.02.97.1.0