Search
Search

Cfap74| Gene Symbol: Cfap74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166029.1 | NP_001159501.1 | 4 - 5 | 685 | 85 | |
| NM_177674.5 | NP_808342.3 | 4 - 5 | 685 | 85 | ||
| GenBank mRNA | AK134858.1 | - | 4 - 5 | 617 | 85 | |
| BC056495.1 | - | 4 - 5 | 617 | 85 | ||
Set Membership: |
|