Search
Search

Atg4c| Gene Symbol: Atg4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145967.1 | NP_001139439.1 | 9 - 10 | 1285 | 138 | |
| NM_175029.3 | NP_778194.3 | 9 - 10 | 1667 | 138 | ||
| GenBank mRNA | AK134838.1 | - | 9 - 10 | 1434 | 138 | |
| BC058981.1 | - | 9 - 10 | 1667 | 6827140 | 138 | |
Set Membership: |
|