| Array Name |
Probe IDs |
Targets |
Target Type |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
711425 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
465828 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
396428 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
AK131931.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
AK150654.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
AK151449.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
AK168911.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
BC025861.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
M74227.1 |
genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
514678 |
NM_008908.4 |
refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
253180 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
859779 |
- |
- |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
167858 |
- |
- |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2054342 |
NR_038009.1 |
mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P2054342 |
AK013046.1 |
mouse_transcriptome |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293834 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293813 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293837 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293805 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293833 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293830 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293809 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293812 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293803 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293799 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293840 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293801 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293815 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00792819 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P00512680 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293838 |
- |
- |
| Agilent Six_ChIP_01_of_03_v2 |
A_68_P20293832 |
- |
- |