Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01260880_g1
          See other AP2S1 GE Assays ›
          Gene Symbol:
          Ap2s1
          Gene Name:
          adaptor-related protein complex 2, sigma 1 subunit
          Gene Aliases:
          AI043088
          Chromosome Location:
          Chr.7: 16738444 - 16749290 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01260880_g1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price 2,694.00
          Your Price
          Online offer:
          2694.0
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Ap2s1
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_198613.2 NP_941015.2 4 - 5 421 82
          GenBank mRNA AK146584.1 - 4 - 5 422 82
          AK186335.1 - 2 - 3 173 82
          AK206960.1 - 2 - 3 173 82
          BC023066.1 - 2 - 3 673 82
          BC052499.1 - 5 - 6 382 3370978 82

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0700000234.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene Ap2s1

          Panther Classification:

          Molecular Function -

          vesicle coat protein

          Gene Ontology Categories:

          Function(s) Process(es)

          transport
          intracellular protein transport
          endocytosis
          protein transport
          vesicle-mediated transport
          molecular_function
          protein transporter activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983044 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983046 - -
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2247852 AK186335.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2247852 AK206960.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2247852 BC023066.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2247852 BC052499.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2247852 AK146584.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 121240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 46833 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 219231 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 781548 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 223240 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 605653 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 46309 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112478 - -
          Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P125549 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2247852 AK186335.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2247852 AK206960.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2247852 BC023066.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2247852 BC052499.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2247852 AK146584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419935 BC052499.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419935 AK146584.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P419935 NM_198613.2 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P128095 AK186335.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P128095 AK206960.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P128095 BC023066.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P128095 BC052499.1 mouse_transcriptome
          Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P128095 AK146584.1 mouse_transcriptome
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983029 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983039 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983040 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983036 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983035 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983052 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983043 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983050 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983031 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983044 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983032 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983054 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983033 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983037 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983030 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983051 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983049 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983027 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983047 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983058 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983056 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983045 - -
          Agilent Mouse Promoter ChIP-on-Chip A) A_68_P24983046 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112478 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06130262 - -
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P125549 - -
          Whole Mouse Genome G4122A A_52_P128095 AK186335.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P128095 AK206960.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P128095 BC023066.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P128095 BC052499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P128095 AK146584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419935 BC052499.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419935 AK146584.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_51_P419935 NM_198613.2 mouse_transcriptome
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P102700 - -
          Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P114464 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P125549 - -
          Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112478 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06063507 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06063492 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06063495 - -
          Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06063508 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983046 - -
          Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P24983044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983076 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983107 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983041 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983027 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983113 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983081 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983108 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983065 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983123 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983009 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983132 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983014 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983095 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983061 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983012 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983039 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983016 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983063 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983126 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983091 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983088 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983127 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983082 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983060 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983074 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983008 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983084 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983047 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983059 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983035 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983037 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983044 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983112 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983024 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983056 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983032 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983134 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983099 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983103 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983050 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983122 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983105 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983071 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983104 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983018 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983129 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983117 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983026 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983051 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983058 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983052 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983054 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983069 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983066 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983111 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983040 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983049 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983079 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983125 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983043 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983033 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983029 - -
          Agilent Mouse Whole Genome ChIP-on-Chip A_68_P24983025 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983066 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063492 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063513 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983043 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983057 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983065 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983035 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983036 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983052 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063504 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983018 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983059 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983022 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983009 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983076 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06604346 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458368 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983069 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06391719 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983031 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063501 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983079 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983037 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063508 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983019 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06303135 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063507 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05906406 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983030 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983038 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063506 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P05906407 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983039 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983073 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983081 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983032 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983056 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06063494 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983072 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06627811 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983075 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06605094 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983040 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983020 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983082 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06118010 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983058 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983008 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P24983033 - -
          Agilent Six_ChIP_02_of_03_v2 A_68_P06458363 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10550329 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550330 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550331 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550327 - -
          Affymetrix Mouse Gene 1.0 ST Array 10550328 - -
          Affymetrix HT MG-430 PM Array Plate 1433612_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10550326 - -
          Mouse Genome 430 2.0 Array 1433612_at - -
          Mouse Genome 430 2.0 Array 1433612_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232910_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 232910_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0700001704.mm.1 TC0700000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001704.mm.1 NM_198613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001704.mm.1 NM_198613.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001702.mm.1 TC0700000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001702.mm.1 NM_198613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001702.mm.1 NM_198613.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001703.mm.1 TC0700000234.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0700001703.mm.1 NM_198613.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0700001703.mm.1 NM_198613.2 Refseq
          Mouse Expression Set 430 1433612_PM_at - -
          Mouse Expression Set 430 1433612_at - -
          Murine Genome U74v2 Set 96639_at - -
          Murine Genome U74v2 Set 96638_at - -
          Murine Genome U74v2 Set 96639_PM_at - -
          Murine Genome U74v2 Set 96638_PM_at - -
          Mouse Genome 430A 2.0 Array 1433612_PM_at - -
          Mouse Genome 430A 2.0 Array 1433612_at - -
          Affymetrix HT Mouse Genome 430A Array 1433612_at - -
          Affymetrix HT Mouse Genome 430A Array 1433612_PM_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 315294 - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 4230402 - -
          Illumina mouse-6 v1.0 expression beadchip 2450524 - -
          Illumina mouse-6 v1.0 expression beadchip 4230402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0004230402 - -
          Illumina mouseRef-8 v1.0 expression beadchip 0002450524 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_1212941 - -
          Illumina mouseRef-8 v1.1 expression beadchip ILMN_2762255 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_2762255 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1212941 - -
          mouse-6 v1.1 (Illumina) scl0232910.6_0-S - -
          mouse-6 v1.1 (Illumina) scl000287.1_1-S - -
          Illumina Sentrix mouse-6.v1 bead array 4230402 - -
          Illumina Sentrix mouse-6.v1 bead array 2450524 - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl0232910.6_0-S - -
          Illumina Sentrix MouseRef-8 Expression BeadChip scl000287.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230402 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (2) 2450524 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2911283 - -
          MouseRef-8 v2 Expression BeadChip ILMN_1212941 - -
          MouseRef-8 v2 Expression BeadChip ILMN_2762255 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2911283 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_2762255 - -
          Illumina MouseWG-6 v2.0 expression beadchip ILMN_1212941 - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl000287.1_1-S - -
          Illumina Sentrix Mouse-6 Expression BeadChip (3) scl0232910.6_0-S - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline