Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033573.1 | NP_001028745.1 | 6 - 7 | 1298 | 129 | |
NM_025883.3 | NP_080159.3 | 6 - 7 | 1182 | 129 | ||
GenBank mRNA | AK010296.1 | - | 6 - 7 | 1177 | 129 | |
AK011373.1 | - | 6 - 7 | 1149 | 129 | ||
AK035172.1 | - | 1261 | 129 | |||
AK038177.1 | - | 1161 | 129 | |||
AK050388.1 | - | 6 - 7 | 1297 | 129 | ||
AK077693.1 | - | 6 - 7 | 1136 | 129 | ||
AK077838.1 | - | 1147 | 129 | |||
AK139707.1 | - | 6 - 7 | 1139 | 129 | ||
AK144128.1 | - | 6 - 7 | 1154 | 129 | ||
AK154741.1 | - | 6 - 7 | 1160 | 129 | ||
AK157494.1 | - | 6 - 7 | 1164 | 129 | ||
AK159304.1 | - | 6 - 7 | 1183 | 129 | ||
AK168497.1 | - | 7 - 8 | 1305 | 129 | ||
BC033317.1 | - | 6 - 7 | 1129 | 4459242 | 129 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |