Search
Search

Pias3| Gene Symbol: Pias3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165949.1 | NP_001159421.1 | 9 - 10 | 1240 | 85 | |
| NM_018812.2 | NP_061282.2 | 10 - 11 | 1135 | 85 | ||
| NM_146135.2 | NP_666247.1 | 9 - 10 | 1277 | 85 | ||
| GenBank mRNA | AF034080.3 | - | 9 - 10 | 1012 | 85 | |
| AK040619.1 | - | 9 - 10 | 1268 | 85 | ||
| AK077551.1 | - | 9 - 10 | 1226 | 85 | ||
| AK085503.1 | - | 9 - 10 | 1212 | 85 | ||
| AK134404.1 | - | 4 - 5 | 415 | 85 | ||
| BC023128.1 | - | 9 - 10 | 1183 | 3585764 | 85 | |
| BC050988.1 | - | 10 - 11 | 1094 | 85 | ||
| BC051252.1 | - | 9 - 10 | 1150 | 6489803 | 85 | |
Set Membership: |
|