Search
Search

Plekha5| Gene Symbol: Plekha5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144920.3 | NP_659169.3 | 6 - 7 | 613 | 55 | |
| GenBank mRNA | AK129423.1 | - | 4 - 5 | 355 | 55 | |
| AK131741.1 | - | 4 - 5 | 384 | 55 | ||
Set Membership: |
|