Search
Search

Snx27| Gene Symbol: Snx27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082484.2 | NP_001075953.1 | 5 - 6 | 1064 | 85 | |
| NM_029721.2 | NP_083997.1 | 5 - 6 | 1064 | 85 | ||
| GenBank mRNA | AK017836.1 | - | 2 - 3 | 135 | 85 | |
| AK122296.1 | - | 5 - 6 | 876 | 85 | ||
| AK147452.1 | - | 5 - 6 | 1051 | 85 | ||
| BC053495.1 | - | 5 - 6 | 876 | 85 | ||
Set Membership: |
|