| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1800011858.mm.1 |
NM_001136181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011858.mm.1 |
NM_001136181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011858.mm.1 |
TC1800001655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011860.mm.1 |
NM_001136181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011860.mm.1 |
NM_001136181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011860.mm.1 |
TC1800001655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011856.mm.1 |
NM_001136181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011856.mm.1 |
NM_001136181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011856.mm.1 |
TC1800001655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011861.mm.1 |
NM_001136181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011861.mm.1 |
NM_001136181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011861.mm.1 |
TC1800001655.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1800011855.mm.1 |
NM_001136181.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011855.mm.1 |
NM_001136181.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1800011855.mm.1 |
TC1800001655.mm.1 |
MTA TC |