Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01262046_m1
          See other KCNG2 GE Assays ›
          Gene Symbol:
          Kcng2
          Gene Name:
          potassium voltage-gated channel, subfamily G, member 2
          Gene Aliases:
          AA591524
          Chromosome Location:
          Chr.18: 80294544 - 80365105 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01262046_m1
          Best Coverage
          Size
          Availability Inventoried
          Catalog # 4331182
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: Kcng2
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NM_001190373.1 NP_001177302.1 3 - 4 952 54

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC1800001657.mm.1

          Set Membership:

          Amplicon length less than or equal to 70 Ion Channel 3' Most Recommended Assay Mus musculus 1700 Inventoried Probe spans exons

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of ion transmembrane transport
          potassium ion transmembrane transport
          delayed rectifier potassium channel activity

          MicroArray Probes:

          Agilent Affymetrix ABI Illumina

          Array Name Probe IDs Targets Target Type
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 154224 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005335 AK158780.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2005335 NM_001190373.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109002 AK158780.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_66_P109002 NM_001190373.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P192085 AK158780.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P192085 NM_001190373.1 mouse_transcriptome
          Array Name Probe IDs Targets Target Type
          Affymetrix HT MG-430 PM Array Plate 1434838_PM_at - -
          Mouse Genome 430 2.0 Array 1446009_PM_at - -
          Mouse Genome 430 2.0 Array 1441695_at - -
          Mouse Genome 430 2.0 Array 1434838_PM_at - -
          Mouse Genome 430 2.0 Array 1446009_at - -
          Mouse Genome 430 2.0 Array 1434838_at - -
          Mouse Genome 430 2.0 Array 1441695_PM_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 240444_at - -
          Affymetrix GeneChip Mouse Genome 430 2.0 Array 240444_PM_at - -
          Affymetrix Mouse Clariom S Array PSR1800011864.mm.1 NM_001190373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011864.mm.1 NM_001190373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011864.mm.1 TC1800001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011865.mm.1 NM_001190373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011865.mm.1 NM_001190373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011865.mm.1 TC1800001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011866.mm.1 NM_001190373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011866.mm.1 NM_001190373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011866.mm.1 TC1800001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011867.mm.1 NM_001190373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011867.mm.1 NM_001190373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011867.mm.1 TC1800001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011868.mm.1 NM_001190373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011868.mm.1 NM_001190373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011868.mm.1 TC1800001657.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR1800011874.mm.1 NM_001190373.1.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011874.mm.1 NM_001190373.1 Refseq
          Affymetrix Mouse Clariom S Array PSR1800011874.mm.1 TC1800001657.mm.1 MTA TC
          Mouse Expression Set 430 1434838_at - -
          Mouse Expression Set 430 1434838_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434838_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1434838_at - -
          Array Name Probe IDs Targets Target Type
          ABI Mouse Genome Survey Microarray 607189 - -
          Array Name Probe IDs Targets Target Type
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216676 - -
          Illumina mouse-6 v1.1 expression beadchip ILMN_1216698 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-fkn7b:80/100.66.79.246:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline