Search
Search

Jakmip2| Gene Symbol: Jakmip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163637.1 | NP_001157109.1 | 7 - 8 | 1694 | 73 | |
| GenBank mRNA | BC094439.1 | - | 6 - 7 | 1256 | 73 | |
Set Membership: |
|