Search
Search

Pcdhgc4| Gene Symbol: Pcdhgc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033582.2 | NP_291060.1 | 1 - 2 | 2909 | 68 | |
| GenBank mRNA | AK139349.1 | - | 1 - 2 | 1647 | 68 | |
| AY013812.1 | - | 1 - 2 | 2455 | 68 | ||
| BC064808.1 | - | 1 - 2 | 2909 | 6825007 | 68 | |
Set Membership: |
|