| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Gene 1.0 ST Array |
10518970 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518973 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518972 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518974 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518971 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10510843 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518968 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518969 |
- |
- |
| Affymetrix Mouse Gene 1.0 ST Array |
10518975 |
- |
- |
| Affymetrix HT MG-430 PM Array Plate |
1456641_PM_at |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10510836 |
- |
- |
| Affymetrix Mouse Gene 1.1 ST Array |
10518967 |
- |
- |
| Mouse Genome 430 2.0 Array |
1456641_PM_at |
- |
- |
| Mouse Genome 430 2.0 Array |
1456641_at |
- |
- |
| Affymetrix Mouse Clariom S Array |
PSR0400034272.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034272.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034272.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034272.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034272.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034273.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034273.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034273.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034273.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034273.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034280.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034280.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034280.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034280.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034280.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034282.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034282.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034282.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034282.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034282.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034278.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034278.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034278.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034278.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034278.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034279.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034279.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034279.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034279.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034279.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034281.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034281.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034281.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034281.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034281.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034275.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034275.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034275.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034275.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034275.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034277.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034277.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034277.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034277.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034277.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034284.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034284.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034284.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034284.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034284.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034291.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034291.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034291.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034291.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034291.mm.1 |
NM_001163722.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034276.mm.1 |
TC0400004112.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400034276.mm.1 |
NM_001163721.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034276.mm.1 |
NM_001163721.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034276.mm.1 |
NM_001163722.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400034276.mm.1 |
NM_001163722.1 |
Refseq |
| Mouse Expression Set 430 |
1456641_at |
- |
- |
| Mouse Expression Set 430 |
1456641_PM_at |
- |
- |
| Murine Genome U74v2 Set |
98458_at |
- |
- |
| Murine Genome U74v2 Set |
98458_PM_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1456641_at |
- |
- |
| Affymetrix HT Mouse Genome 430B Array |
1456641_PM_at |
- |
- |