Search
Search

Slc25a33| Gene Symbol: Slc25a33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027460.2 | NP_081736.2 | 1 - 2 | 273 | 65 | |
| GenBank mRNA | AK017628.1 | - | 1 - 2 | 185 | 65 | |
| AK157423.1 | - | 1 - 2 | 274 | 65 | ||
| BC011293.1 | - | 1 - 2 | 248 | 3989313 | 65 | |
| BQ950692.1 | - | 1 - 2 | 208 | 65 | ||
| CK795124.1 | - | 1 - 2 | 282 | 65 | ||
Set Membership: |
|