Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          • Home
          • › Search Tool
          • › Search Results
          • › Mm01264015_g1
          See other E130114P18RIK GE Assays ›
          Gene Symbol:
          E130114P18Rik
          Gene Name:
          RIKEN cDNA E130114P18 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 97567875 - 97584591 on Build GRCm38
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          Assay ID Mm01264015_g1
          Best Coverage
          Size
          Availability Made To Order
          Catalog # 4351372
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • Assay Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          Assay Details

          Gene Symbol: E130114P18Rik
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
          RefSeq NR_015513.2 2 - 3 138 76
          GenBank mRNA AK053617.1 - 298 76
          AK082601.1 - 2 - 3 140 76
          AK086199.1 - 474 76
          AK086602.1 - 2 - 3 184 76
          BC048947.1 - 3 - 4 257 76
          Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
          GenBankmRNA NM_177023.1 - 474 Chr. - - [-] on Build 76

          Back To Top

          More Information


          Updated Mapping Information

          This assay targets the same location as MTA transcript cluster(s) TC0400003106.mm.1

          Set Membership:

          Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons

          Matching Ambion siRNAs:

          Click here to find siRNAs for Gene E130114P18Rik

          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          MicroArray Probes:

          Agilent Affymetrix Illumina

          Array Name Probe IDs Targets Target Type
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976589 AK082601.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976589 AK086199.1 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976589 NR_015513.2 mouse_transcriptome
          Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P1976589 AK053617.1 mouse_transcriptome
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 70835 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 141131 - -
          Agilent SurePrint G3 Mouse CGH Microarray 1x1M 33467 - -
          Agilent SurePrint G3 Mouse CGH Microarray 4x180K 96515 - -
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976589 AK082601.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976589 AK086199.1 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976589 NR_015513.2 mouse_transcriptome
          SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P1976589 AK053617.1 mouse_transcriptome
          Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P05327392 - -
          Whole Mouse Genome G4122A A_52_P376039 AK053617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337910 AK086199.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337910 AK053617.1 mouse_transcriptome
          Whole Mouse Genome G4122A A_52_P337910 BC048947.1 mouse_transcriptome
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132882 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132876 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132880 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132881 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132877 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132873 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132872 - -
          Agilent Six_ChIP_01_of_03_v2 A_68_P23132878 - -
          Array Name Probe IDs Targets Target Type
          Affymetrix Mouse Gene 1.0 ST Array 10514566 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514567 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514563 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514564 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514565 - -
          Affymetrix Mouse Gene 1.0 ST Array 10514562 - -
          Affymetrix HT MG-430 PM Array Plate 1435765_PM_at - -
          Affymetrix Mouse Gene 1.1 ST Array 10514561 - -
          Mouse Genome 430 2.0 Array 1435765_PM_at - -
          Mouse Genome 430 2.0 Array 1435765_at - -
          Mouse Genome 430 2.0 Array 1438135_at - -
          Mouse Genome 430 2.0 Array 1438135_PM_at - -
          Affymetrix Mouse Clariom S Array PSR0400024855.mm.1 TC0400003106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024855.mm.1 NR_015513.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024855.mm.1 NR_015513.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024854.mm.1 TC0400003106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024854.mm.1 NR_015513.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024854.mm.1 NR_015513.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024864.mm.1 TC0400003106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024864.mm.1 NR_015513.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024864.mm.1 NR_015513.2 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024857.mm.1 TC0400003106.mm.1 MTA TC
          Affymetrix Mouse Clariom S Array PSR0400024857.mm.1 NR_015513.2.1 Refseq
          Affymetrix Mouse Clariom S Array PSR0400024857.mm.1 NR_015513.2 Refseq
          Mouse Expression Set 430 1438135_PM_at - -
          Mouse Expression Set 430 1438135_at - -
          Mouse Expression Set 430 1435765_PM_at - -
          Mouse Expression Set 430 1435765_at - -
          Murine Genome U74v2 Set 135145_PM_r_at - -
          Murine Genome U74v2 Set 135145_r_at - -
          Affymetrix HT Mouse Genome 430B Array 1435765_PM_at - -
          Affymetrix HT Mouse Genome 430B Array 1438135_at - -
          Affymetrix HT Mouse Genome 430B Array 1435765_at - -
          Affymetrix HT Mouse Genome 430B Array 1438135_PM_at - -
          Array Name Probe IDs Targets Target Type
          MouseWG-6 v2 Expression BeadChip 450131 - -

          Back To Top

          Related Products

          • TaqMan® Fast Advanced Master Mix
          • SuperScript® VILO™ cDNA Synthesis Kit
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
          git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.45.0-Offline