Search
Search

Pcdh1| Gene Symbol: Pcdh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029357.3 | NP_083633.2 | 2 - 3 | 1067 | 79 | |
| GenBank mRNA | AY861431.1 | - | 2 - 3 | 978 | 79 | |
| BC024121.1 | - | 1 - 2 | 618 | 79 | ||
| BC039218.1 | - | 1 - 2 | 139 | 79 | ||
| BC040402.1 | - | 2 - 3 | 1067 | 4162978 | 79 | |
Set Membership: |
|