Search
Search

Zfp507| Gene Symbol: Zfp507 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177739.3 | NP_808407.2 | 3 - 4 | 2340 | 70 | |
| GenBank mRNA | AK129283.1 | - | 3 - 4 | 2308 | 70 | |
| AK162510.1 | - | 1 - 2 | 520 | 70 | ||
| BC050041.1 | - | 3 - 4 | 2329 | 30006851 | 70 | |
Set Membership: |
|