Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
Search Tool
›
Search Results
› Mm01264678_g1
See other 2210420H20RIK GE Assays ›
Gene Symbol:
2210420H20Rik
Gene Name:
RIKEN cDNA 2210420H20 gene
Gene Aliases:
-
Chromosome Location:
Chr.18: 82754505 - 82759039 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID
Mm01264678_g1
Best Coverage
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: 2210420H20Rik
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NR_045389.1
3 - 4
563
63
GenBank mRNA
AK008989.1
-
3 - 4
565
63
Back To Top
More Information
Updated Mapping Information
This assay targets the same location as MTA transcript cluster(s) TC1800001683.mm.1
Set Membership:
Amplicon length less than or equal to 70
3' Most
Detect Genomic DNA
Recommended Assay
Mus musculus
1700
Probe spans exons
MicroArray Probes:
Agilent
Affymetrix
ABI
Array Name
Probe IDs
Targets
Target Type
Agilent SurePrint G3 Mouse CGH Microarray 1x1M
684881
-
-
Agilent SurePrint G3 Mouse CGH Microarray 4x180K
79332
-
-
Agilent Mouse Genome CGH Microarray 244A (G4415A)
A_67_P03528018
-
-
Agilent Mouse Developmental Microarray 44K G2519A Option 002
A_66_P138712
-
-
Agilent Mouse Genome CGH Microarray 105A (G4416A)
A_67_P03528018
-
-
Array Name
Probe IDs
Targets
Target Type
Affymetrix Mouse Clariom S Array
PSR1800012054.mm.1
NR_045389.1.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1800012054.mm.1
NR_045389.1
Refseq
Affymetrix Mouse Clariom S Array
PSR1800012054.mm.1
TC1800001683.mm.1
MTA TC
Array Name
Probe IDs
Targets
Target Type
ABI Mouse Genome Survey Microarray
835461
-
-
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-64dd947b88-tr4x4:80/100.66.75.98:80.
git-commit: 0f46c0ba67a87c24f5ac662a3edafcaba07cd08c
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.45.0-Offline