Search
Search

Prkaa2| Gene Symbol: Prkaa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178143.2 | NP_835279.2 | 6 - 7 | 888 | 64 | |
| GenBank mRNA | AK044030.1 | - | 6 - 7 | 724 | 64 | |
| AK134573.1 | - | 6 - 7 | 746 | 64 | ||
| BC138565.1 | - | 6 - 7 | 897 | 8861585 | 64 | |
| BC138566.1 | - | 6 - 7 | 897 | 8861586 | 64 | |
| FJ588504.1 | - | 5 - 6 | 646 | 64 | ||
Set Membership: |
|