Search
Search

8430419L09Rik| Gene Symbol: 8430419L09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028982.4 | NP_083258.2 | 9 - 10 | 1453 | 71 | |
| GenBank mRNA | AK018428.1 | - | 9 - 10 | 1397 | 71 | |
| AK039401.1 | - | 9 - 10 | 1389 | 71 | ||
| AK220395.1 | - | 8 - 9 | 1103 | 71 | ||
| BC003918.1 | - | 1 - 2 | 52 | 71 | ||
| BC038002.1 | - | 8 - 9 | 1114 | 71 | ||
| BC079676.1 | - | 9 - 10 | 1380 | 71 | ||
| BC137674.1 | - | 9 - 10 | 1432 | 8860693 | 71 | |
| BC137676.1 | - | 9 - 10 | 1432 | 8860695 | 71 | |
Set Membership: |
|